missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(3-methoxyphenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61581.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416626

  • £86.92 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(3-methoxyphenylcarbamoyl)benzeneboronic acid
349.98
MFCD26143292

Specifications

3-Bromo-5-(3-methoxyphenylcarbamoyl)benzeneboronic acid
1g
349.98
Solid
97%
MFCD26143292
Videos
SDS