missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(2-ethylphenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61124.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456286

  • £95.61 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(2-ethylphenylcarbamoyl)benzeneboronic acid
349.01
MFCD26143276

Specifications

3-Bromo-5-(2-ethylphenylcarbamoyl)benzeneboronic acid
97%
349.01
Solid
1g
MFCD26143276
Videos
SDS