missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61491.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406556

  • £95.61 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid
389.85
MFCD26143287

Specifications

3-Bromo-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid
97%
MFCD26143287
Solid
389.85
1g
Videos
SDS