missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitrobenzotrifluoride, 95%, Thermo Scientific™

Catalog Number 15431559
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
132839-58-8
C7H3BrF3NO2
270.005
MFCD09998160
DJOLBPUSSIOMKU-UHFFFAOYSA-N
2-bromo-1-nitro-4-trifluoromethyl benzene, 3-bromo-4-nitrobenzotrifluoride, 2-bromo-4-trifluoromethyl nitrobenzene, benzene, 2-bromo-1-nitro-4-trifluoromethyl, acmc-209bq5, ksc496a5t, 2-bromo-1-nitro-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethylnitrobenzene
14790878
2-bromo-1-nitro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]
This item is not returnable. View return policy
132839-58-8
C7H3BrF3NO2
270.005
MFCD09998160
DJOLBPUSSIOMKU-UHFFFAOYSA-N
2-bromo-1-nitro-4-trifluoromethyl benzene, 3-bromo-4-nitrobenzotrifluoride, 2-bromo-4-trifluoromethyl nitrobenzene, benzene, 2-bromo-1-nitro-4-trifluoromethyl, acmc-209bq5, ksc496a5t, 2-bromo-1-nitro-4-trifluoromethyl-benzene, 2-bromo-4-trifluoromethylnitrobenzene
14790878
2-bromo-1-nitro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Formula Weight 270.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-nitrobenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'