missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylbenzotrifluoride, 97%, ACROS Organics™

Brand:  Acros Organics 446680250

Code : 29

Additional Details : CAS Number : 66417-30-9 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11994551

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Bromo-4-methylbenzotrifluoride
97%
C8H6BrF3
239.03

Specifications

3-Bromo-4-methylbenzotrifluoride
25g
C8H6BrF3
66417-30-9
97%
239.03
Videos
SDS