missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-methylbenzoic acid, 98+%, Thermo Scientific Chemicals
23.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Melting Point | 206°C to 211°C |
Odor | Odorless |
Quantity | 5 g |
Beilstein | 1936510 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 215.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Bromo-4-methylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'