missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7055659
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
82257-09-8
C6H6BrNO
188.024
MFCD05664024
DSFJKFHIMBVWAV-UHFFFAOYSA-N
642827
3-bromo-4-methoxypyridine
COC1=C(C=NC=C1)Br
This item is not returnable. View return policy
82257-09-8
C6H6BrNO
188.024
MFCD05664024
DSFJKFHIMBVWAV-UHFFFAOYSA-N
642827
3-bromo-4-methoxypyridine
COC1=C(C=NC=C1)Br

3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Beilstein 1423610
Synonym 3-bromo-4-methoxy-pyridine, pyridine, 3-bromo-4-methoxy, pyridine,3-bromo-4-methoxy, pubchem6686, 4-methoxy-3-bromopyridine, acmc-209po0, abbypharma ap-17-5958
Solubility Information Slightly soluble in water (9.8g/L).
Formula Weight 188.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methoxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'