missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog Number 15458268
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
27407-11-0
C6H4BrFO
190.999
MFCD03425884
QWTULQLVGNZMLF-UHFFFAOYSA-N
3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol
2783381
3-bromo-4-fluorophenol
C1=CC(=C(C=C1O)Br)F
This item is not returnable. View return policy
27407-11-0
C6H4BrFO
190.999
MFCD03425884
QWTULQLVGNZMLF-UHFFFAOYSA-N
3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol
2783381
3-bromo-4-fluorophenol
C1=CC(=C(C=C1O)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Green
Quantity 1 g
UN Number UN2811
Formula Weight 191
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 3-Bromo-4-fluorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'