missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive. Moisture sensitive. Store under Argon.
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive. Moisture sensitive. Store under Argon.
Specifications
Melting Point | 41°C |
Color | White |
Quantity | 25 g |
UN Number | UN3261 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 267.92 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 3-Bromo-4-fluorobenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'