missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-cyanopyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 91°C to 94°C |
Assay Percent Range | 96% min. (GC) |
Quantity | 5 g |
Synonym | 3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo |
Formula Weight | 183.01 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-4-cyanopyridine |
Safety and Handling
Product Identifier
- 3-Bromo-4-cyanopyridine
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'