missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4089954
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42860-10-6
C7H4BrClO2
235.46
MFCD00079706
NLEPZGNUPNMRGF-UHFFFAOYSA-N
3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
2735544
3-bromo-4-chlorobenzoic acid
OC(=O)C1=CC(Br)=C(Cl)C=C1
This item is not returnable. View return policy
42860-10-6
C7H4BrClO2
235.46
MFCD00079706
NLEPZGNUPNMRGF-UHFFFAOYSA-N
3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
2735544
3-bromo-4-chlorobenzoic acid
OC(=O)C1=CC(Br)=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 235.47
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-chlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'