missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7047445
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42860-10-6
C7H4BrClO2
235.46
MFCD00079706
NLEPZGNUPNMRGF-UHFFFAOYSA-N
3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
2735544
3-bromo-4-chlorobenzoic acid
OC(=O)C1=CC(Br)=C(Cl)C=C1
This item is not returnable. View return policy
42860-10-6
C7H4BrClO2
235.46
MFCD00079706
NLEPZGNUPNMRGF-UHFFFAOYSA-N
3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
2735544
3-bromo-4-chlorobenzoic acid
OC(=O)C1=CC(Br)=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 218°C to 222°C
Quantity 1 g
Beilstein 2438796
Formula Weight 235.47
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'