missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-3-buten-1-ol, 97+%, Thermo Scientific Chemicals

Catalog Number p-3735690
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
76334-36-6
C4H7BrO
151
MFCD00154041
RTKMFQOHBDVEBC-UHFFFAOYSA-N
3-bromo-3-buten-1-ol, 3-buten-1-ol, 3-bromo, 3bb, zlchem 79, acmc-20an90, 3-bromanylbut-3-en-1-ol
533975
3-bromobut-3-en-1-ol
C=C(CCO)Br
This item is not returnable. View return policy
76334-36-6
C4H7BrO
151
MFCD00154041
RTKMFQOHBDVEBC-UHFFFAOYSA-N
3-bromo-3-buten-1-ol, 3-buten-1-ol, 3-bromo, 3bb, zlchem 79, acmc-20an90, 3-bromanylbut-3-en-1-ol
533975
3-bromobut-3-en-1-ol
C=C(CCO)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Brown
Density 1.5220g/mL
Boiling Point 175.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(Br)CH2CH2OH
Refractive Index 1.4949
Quantity 10 g
Beilstein 01, IV, 2100
Specific Gravity 1.522
Formula Weight 151
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-Bromo-3-buten-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-3-buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'