missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylimidazo[1,2-a]pyridine, 97%, Maybridge

Brand:  Maybridge CC21710DE

Code : 31

Additional Details : CAS Number : 4805-70-3 Weight : 0.05500kg

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10461134

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-Bromo-2-methylimidazo[1, 2-a]pyridine
97%
211.06
97%
C8H7BrN2

Specifications

3-Bromo-2-methylimidazo[1, 2-a]pyridine
Amber Glass Bottle
97%
C8H7BrN2
4805-70-3
5g
97%
211.06
Videos
SDS