missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 98%

Brand:  Alfa Aesar™ H64713.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422029

  • £35.19 / 5g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine
76041-73-1
242
MFCD02691223

Specifications

3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine
170° to 171°C
98%
242
76041-73-1
Solid
5g
MFCD02691223
Videos
SDS