missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number 10666440
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
76041-73-1
C6H3BrF3NO
241.99
MFCD02691223
UWHKLLYQUQPOQK-UHFFFAOYSA-N
3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531
13491775
3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)Br
This item is not returnable. View return policy
76041-73-1
C6H3BrF3NO
241.99
MFCD02691223
UWHKLLYQUQPOQK-UHFFFAOYSA-N
3-bromo-5-trifluoromethyl pyridin-2-ol, 3-bromo-2-hydroxy-5-trifluoromethyl pyridine, 3-bromo-2-hydroxy-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl pyridin-2 1h-one, 3-bromo-5-trifluoromethyl pyridine-2-ol, 3-bromo-5-trifluoromethyl-2-pyridinol, abbypharma ap-12-5330, 2 1h-pyridinone, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl-1h-pyridin-2-one, pubchem14531
13491775
3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)Br
CAS Min % 96.0
CAS Max % 100.0
Melting Point 170.1°C to 171.2°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 241.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'