missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15492119
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
352535-97-8
C6H5BBrFO2
218.816
MFCD05664228
QCLOSORNRRMFIA-UHFFFAOYSA-N
3-bromo-2-fluorophenyl boronic acid, 2-fluoro-3-bromophenylboronic acid, boronic acid,b-3-bromo-2-fluorophenyl, boronic acid, b-3-bromo-2-fluorophenyl, 3-bromo-2-fluorophenylboronicacid, acmc-1aijv, 3-bromo-2-fluoro-phenylboronic acid, 3-bromo-2-fluorobenzeneboronic acid, 3-bromanyl-2-fluoranyl-phenyl boronic acid, contains varying amounts of anhydride
16217170
(3-bromo-2-fluorophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)F)(O)O
This item is not returnable. View return policy
352535-97-8
C6H5BBrFO2
218.816
MFCD05664228
QCLOSORNRRMFIA-UHFFFAOYSA-N
3-bromo-2-fluorophenyl boronic acid, 2-fluoro-3-bromophenylboronic acid, boronic acid,b-3-bromo-2-fluorophenyl, boronic acid, b-3-bromo-2-fluorophenyl, 3-bromo-2-fluorophenylboronicacid, acmc-1aijv, 3-bromo-2-fluoro-phenylboronic acid, 3-bromo-2-fluorobenzeneboronic acid, 3-bromanyl-2-fluoranyl-phenyl boronic acid, contains varying amounts of anhydride
16217170
(3-bromo-2-fluorophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 225°C to 227°C (literature)
Quantity 1 g
Beilstein 14006009
Formula Weight 218.82
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'