missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
22.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It can be used as pharmaceutical raw material and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
It can be used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications
Melting Point | 43°C to 46°C |
Color | White |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
Beilstein | 13904499 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 203.01 |
Percent Purity | 97% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'