missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15461939
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
149947-15-9
C7H4BrFO
203.01
MFCD09027089
OUAZPCKRSSEQKB-UHFFFAOYSA-N
3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
22019361
3-bromo-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)F)C=O
This item is not returnable. View return policy
149947-15-9
C7H4BrFO
203.01
MFCD09027089
OUAZPCKRSSEQKB-UHFFFAOYSA-N
3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
22019361
3-bromo-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)F)C=O

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 43°C to 46°C
Color White
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 13904499
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'