missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-fluoroacetophenone, 96%

Brand:  Alfa Aesar™ H62708.06

Code : NEW

Additional Details : CAS Number : 161957-61-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419198

  • £483.30 / 5g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2-fluoroacetophenone
161957-61-5
217.04

Specifications

3-Bromo-2-fluoroacetophenone
Solid
5g
161957-61-5
96%
217.04
Videos
SDS