missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-fluoro-5-methylpyridine, 98%

Brand:  Alfa Aesar™ H61104.14

Code : NEW

Additional Details : CAS Number : 17282-01-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15417608

  • £182.32 / 25g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2-fluoro-5-methylpyridine
17282-01-8
190.01
MFCD03095305

Specifications

3-Bromo-2-fluoro-5-methylpyridine
1.600g/mL
Solid
162° to 164°C (750mmHg)
190.01
17282-01-8
58° to 60°C
98%
25g
MFCD03095305
Videos
SDS