missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloropyridine, 98%, Acros Organics™

Brand:  Acros Organics 360650250

Code : 29

Additional Details : CAS Number : 52200-48-3 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 10556820

  • £75.50 / 25g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

3-Bromo-2-chloropyridine
97.5
98%
C5H3BrClN
52200-48-3
100.0
97%
192.44

Specifications

3-Bromo-2-chloropyridine
97.5
Authentic
Crystalline Powder, Crystals or Flakes or Solid
97%
192.44
57.9° to 58.3°C
25g
52200-48-3
100.0
Glass bottle
98%
C5H3BrClN
White to yellow
97.0°C (1.3 kPa)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Bromo-2-chloropyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS