missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-chloro-6-fluorotoluene, 97%

Brand:  Alfa Aesar™ H63276.03

Code : NEW

Additional Details : CAS Number : 203302-92-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419898

  • £10.07 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2-chloro-6-fluorotoluene
223.47
1-Bromo-2-chloro-4-fluoro-3-methylbenzene
203302-92-5
MFCD00236591

Specifications

3-Bromo-2-chloro-6-fluorotoluene
1g
97%
MFCD00236591
203302-92-5
Solid
223.47
1-Bromo-2-chloro-4-fluoro-3-methylbenzene
Videos
SDS