missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,5-diaminopyridine, 99%, Thermo Scientific Chemicals

Catalog Number 10610893
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
896160-69-3
C5H6BrN3
188.03
MFCD03427651
YJENCMIONVUOCW-UHFFFAOYSA-N
2,5-diamino-3-bromopyridine, 3-bromo-2,5-diaminopyridine, 2,5-diamino-3-bormopyridine, 2,5-pyridinediamine, 3-bromo, pubchem5490, acmc-1c3l9, 3-bromanylpyridine-2,5-diamine, abbypharma ap-31-4166
2734413
3-bromopyridine-2,5-diamine
NC1=CC(Br)=C(N)N=C1
This item is not returnable. View return policy
896160-69-3
C5H6BrN3
188.03
MFCD03427651
YJENCMIONVUOCW-UHFFFAOYSA-N
2,5-diamino-3-bromopyridine, 3-bromo-2,5-diaminopyridine, 2,5-diamino-3-bormopyridine, 2,5-pyridinediamine, 3-bromo, pubchem5490, acmc-1c3l9, 3-bromanylpyridine-2,5-diamine, abbypharma ap-31-4166
2734413
3-bromopyridine-2,5-diamine
NC1=CC(Br)=C(N)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 117°C to 122°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 188
Percent Purity 99%
Chemical Name or Material 3-Bromo-2, 5-diaminopyridine
compliance-icons
Product Identifier
  • 3-Bromo-2,5-diaminopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'