missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2,4,5,6-tetrafluorobenzoyl chloride, 98%

Brand:  Alfa Aesar™ L20152.14

Code : NEW

Additional Details : CAS Number : 292621-46-6 Weight : 0.02500kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11358956

  • £381.90 / 25g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2,4,5,6-tetrafluorobenzoyl chloride
C7BrClF4O
MFCD05664064
292621-46-6
291.43
UN3265

Specifications

3-Bromo-2,4,5,6-tetrafluorobenzoyl chloride
98%
291.43
UN3265
25g
292621-46-6
C7BrClF4O
MFCD05664064
142° to 144°C (80mmHg)
>100°C (212°F)
Videos
SDS