missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-methylpyrrolidine, 97%, Thermo Scientific™

Catalog Number 15492709
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10603-45-9
C5H10BrN
164.046
MFCD12755877
ZHUYTHOKJZSZQJ-UHFFFAOYSA-N
3-bromo-1-methyl-pyrrolidine, pyrrolidine, 3-bromo-1-methyl, 3-bromo-1-methylpyrrolidine
13726279
3-bromo-1-methylpyrrolidine
CN1CCC(C1)Br
This item is not returnable. View return policy
10603-45-9
C5H10BrN
164.046
MFCD12755877
ZHUYTHOKJZSZQJ-UHFFFAOYSA-N
3-bromo-1-methyl-pyrrolidine, pyrrolidine, 3-bromo-1-methyl, 3-bromo-1-methylpyrrolidine
13726279
3-bromo-1-methylpyrrolidine
CN1CCC(C1)Br
Color Yellow to Brown
Density 1.361 g/mL
Flash Point 54°C (129°F)
Refractive Index 1.491
Quantity 1 g
UN Number UN2733
Sensitivity Moisture sensitive
Formula Weight 164.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-methylpyrrolidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'