missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-1-methylpyrrolidine, 97%

Brand:  Alfa Aesar™ H66250.MD

Code : NEW

Additional Details : CAS Number : 10603-45-9 Weight : 0.00025kg Transport : UN number : 2733 Chem class : 3 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15412719

  • £30.20 / 250mg


Estimated Shipment date
from Supplier 20-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-1-methylpyrrolidine
164.04
UN2733
10603-45-9
MFCD12755877

Specifications

3-Bromo-1-methylpyrrolidine
1.4910
250mg
97%
Moisture Sensitive
MFCD12755877
10603-45-9
1.361g/mL
Liquid
Yellow to brown
164.04
UN2733
Videos
SDS