missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-1-methylpyrrolidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Color | Yellow to Brown |
Density | 1.361 g/mL |
Flash Point | 54°C (129°F) |
Refractive Index | 1.491 |
Quantity | 5 g |
UN Number | UN2733 |
Sensitivity | Moisture sensitive |
Formula Weight | 164.04 |
Percent Purity | 97% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'