missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-benzofuran, 97%, Maybridge

Brand:  Maybridge CC27510DE

Code : 31

Additional Details : CAS Number : 59214-70-9 Weight : 0.05500kg

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10694965

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Bromo-1-benzofuran
97%
197.03
97%
C8H5BrO

Specifications

3-Bromo-1-benzofuran
Amber Glass Bottle
97%
C8H5BrO
59214-70-9
5g
97%
197.03
Videos
SDS