missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyaniline, 98%, Thermo Scientific Chemicals

Catalog Number 15412038
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1484-26-0
C13H13NO
199.253
MFCD00007784
IGPFOKFDBICQMC-UHFFFAOYSA-N
3-benzyloxyaniline, 3-benzyloxy aniline, benzenamine, 3-phenylmethoxy, unii-iab1vda972, 3-benzyloxy-phenylamine, iab1vda972, 3-phenylmethyl oxy aniline, 3-phenylmethoxy phenylamine, 3-benzyloxy benzenamine, 3-benyloxyaniline
92892
3-phenylmethoxyaniline
C1=CC=C(C=C1)COC2=CC=CC(=C2)N
This item is not returnable. View return policy
1484-26-0
C13H13NO
199.253
MFCD00007784
IGPFOKFDBICQMC-UHFFFAOYSA-N
3-benzyloxyaniline, 3-benzyloxy aniline, benzenamine, 3-phenylmethoxy, unii-iab1vda972, 3-benzyloxy-phenylamine, iab1vda972, 3-phenylmethyl oxy aniline, 3-phenylmethoxy phenylamine, 3-benzyloxy benzenamine, 3-benyloxyaniline
92892
3-phenylmethoxyaniline
C1=CC=C(C=C1)COC2=CC=CC(=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 63°C to 67°C
Color Brown
Quantity 1 g
Formula Weight 199.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Benzyloxyaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'