missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-2699165
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
25 g
This item is not returnable. View return policy
156682-54-1
C13H13BO3
228.05
MFCD02093063
WIJNYNBSPQMJGO-UHFFFAOYSA-N
3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
2734313
[3-(benzyloxy)phenyl]boronic acid
OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1
This item is not returnable. View return policy
156682-54-1
C13H13BO3
228.05
MFCD02093063
WIJNYNBSPQMJGO-UHFFFAOYSA-N
3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
2734313
[3-(benzyloxy)phenyl]boronic acid
OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 141.0°C to 142.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 228.05
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Benzyloxybenzeneboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzyloxybenzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'