missing translation for 'onlineSavingsMsg'
Learn More

3-{[(Benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3540977
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
900799-69-1
C15H13NO5
287.271
MFCD09817564
DIPYTSQSLOJNFI-UHFFFAOYSA-N
3-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxy carbonyl amino-5-hydroxybenzoic acid, 3-n-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxycarbonylamino-5-hydroxy-benzoic acid, benzoicacid, 3-hydroxy-5-phenylmethoxy carbonyl amino, benzoicacid,3-hydroxy-5-phenylmethoxy carbonyl amino
24229785
3-hydroxy-5-(phenylmethoxycarbonylamino)benzoic acid
C1=CC=C(C=C1)COC(=O)NC2=CC(=CC(=C2)C(=O)O)O
This item is not returnable. View return policy
900799-69-1
C15H13NO5
287.271
MFCD09817564
DIPYTSQSLOJNFI-UHFFFAOYSA-N
3-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxy carbonyl amino-5-hydroxybenzoic acid, 3-n-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxycarbonylamino-5-hydroxy-benzoic acid, benzoicacid, 3-hydroxy-5-phenylmethoxy carbonyl amino, benzoicacid,3-hydroxy-5-phenylmethoxy carbonyl amino
24229785
3-hydroxy-5-(phenylmethoxycarbonylamino)benzoic acid
C1=CC=C(C=C1)COC(=O)NC2=CC(=CC(=C2)C(=O)O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 287.27
Percent Purity 97%
Chemical Name or Material 3-{[(Benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'