missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-1-propanol, 97%, Thermo Scientific Chemicals

Catalog Number p-4228366
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4799-68-2
C10H14O2
166.22
MFCD00029659
FUCYABRIJPUVAT-UHFFFAOYSA-N
3-benzyloxy-1-propanol, 3-benzyloxy propan-1-ol, 3-benzyloxypropanol, 1-propanol, 3-phenylmethoxy, 3-benzyloxy-propan-1-ol, 1-propanol, 3-benzyloxy, 3-benzyloxypropan-1-ol, 3-benzyloxy-propane-1-ol, 3-benzyl-oxy propan-1-ol
347971
OCCCOCC1=CC=CC=C1
This item is not returnable. View return policy
4799-68-2
C10H14O2
166.22
MFCD00029659
FUCYABRIJPUVAT-UHFFFAOYSA-N
3-benzyloxy-1-propanol, 3-benzyloxy propan-1-ol, 3-benzyloxypropanol, 1-propanol, 3-phenylmethoxy, 3-benzyloxy-propan-1-ol, 1-propanol, 3-benzyloxy, 3-benzyloxypropan-1-ol, 3-benzyloxy-propane-1-ol, 3-benzyl-oxy propan-1-ol
347971
OCCCOCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0490g/mL
Boiling Point 111°C to 114°C (2.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.049
IUPAC Name 3-phenylmethoxypropan-1-ol
Formula Weight 166.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Benzyloxy-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzyloxy-1-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'