missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Benzyl-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63800.06

Code : NEW

Additional Details : CAS Number : 93286-13-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470689

  • £331.78 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Benzyl-1-phenyl-2-thiohydantoin
282.36
15205524
93286-13-6
MFCD27665237
3-Benzyl-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-Benzyl-1-phenyl-2-thiohydantoin
Solid
282.36
15205524
3-Benzyl-1-phenyl-2-thioxo-4-imidazolidinone
93286-13-6
95%
MFCD27665237
5g
Videos
SDS