missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylbenzoic acid, 99%, Acros Organics™

Brand:  Acros Organics 339090050

Code : NEW

Additional Details : CAS Number : 579-18-0 Weight : 0.01000kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10481373

  • £56.60 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket


Specifications

Chemical Identifiers

3-Benzoylbenzoic acid
579-18-0
100.0
99%
C6H5COC6H4COOH
98%
97.5
99%
C14H10O3
226.23

Specifications

3-Benzoylbenzoic acid
579-18-0
100.0
99%
C14H10O3
226.23
Glass bottle
161.0° to 166.5°C
98%
97.5
Powder
99%
C6H5COC6H4COOH
Authentic
White to light beige
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Benzoylbenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS