missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 3-Azido-7-hydroxycoumarin
GREENER_CHOICE

Product Code. 18707526
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18707526

Brand: Tocris Bioscience™ 7664/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Click-reactive fluorescent dye

3-Azido-7-hydroxycoumarin is a water-soluble fluorogenic fluorescent dye. It emits intense fluorescence only after 'click' reaction when it forms triazoles at the site of alkyne modified biomolecules. Excitation and emission maxima are 404 nm and 477 nm respectively, after click reaction.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Azido-7-hydroxy-2 H-1-benzopyran-2-one
CAS 817638-68-9
Quantity 5 mg
Target Fluorescent Dyes
Molecular Formula C9H5N3O3
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.