missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Azabicyclo[3.3.0]octane hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Melting Point | 116.0°C to 118.5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5 to max. 101.5% min. (Argentometry) |
Quantity | 5 g |
Formula Weight | 147.65 |
Percent Purity | 99% |
Chemical Name or Material | 3-Azabicyclo[3.3.0]octane hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'