missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Employed as an important intermediate for syntheses material and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as an important intermediate for syntheses material and pharmaceutical.
Solubility
Soluble in acetone, ethanol and methanol.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Employed as an important intermediate for syntheses material and pharmaceutical.
Solubility
Soluble in acetone, ethanol and methanol.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 218°C to 222°C |
Color | Brown |
Quantity | 5 g |
Beilstein | 3600 |
Solubility Information | Soluble in acetone,ethanol and methanol. |
Formula Weight | 138.13 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 3-Aminopyridine-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'