missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15442259
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1462-86-8
C6H6N2O2
138.126
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N
This item is not returnable. View return policy
1462-86-8
C6H6N2O2
138.126
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N

Employed as an important intermediate for syntheses material and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for syntheses material and pharmaceutical.

Solubility
Soluble in acetone, ethanol and methanol.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 218°C to 222°C
Color Brown
Quantity 25 g
Beilstein 3600
Solubility Information Soluble in acetone,ethanol and methanol.
Formula Weight 138.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Aminopyridine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'