missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228341
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1462-86-8
C6H6N2O2
138.13
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N
This item is not returnable. View return policy
1462-86-8
C6H6N2O2
138.13
MFCD00090153
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
73836
3-aminopyridine-2-carboxylic acid
C1=CC(=C(N=C1)C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 200°C to 201°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 138.13
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3-Aminopyridine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopyridine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'