missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltriethoxysilane, 99%, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4228340
40.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
919-30-2
C9H23NO3Si
221.37
MFCD00008207,MFCD01324904
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
919-30-2
C9H23NO3Si
221.37
MFCD00008207,MFCD01324904
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Density 0.9420g/mL
Boiling Point 217.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula H2N(CH2)3Si(OC2H5)3
Refractive Index 1.4200 to 1.4220
Quantity 100 mL
Specific Gravity 0.942
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Aminopropyltriethoxysilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'