missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltriethoxysilane, 99%, Thermo Scientific Chemicals

Catalog Number p-4510201
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
919-30-2
C9H23NO3Si
221.37
MFCD00008207,MFCD01324904
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
919-30-2
C9H23NO3Si
221.37
MFCD00008207,MFCD01324904
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70.0°C
Density 0.9420g/mL
Boiling Point 217.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)3Si(OC2H5)3
Refractive Index 1.4200 to 1.4220
Quantity 5 g
Beilstein 04,IV,4273
Specific Gravity 0.942
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'