missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyldimethylmethoxysilane, 97%, ACROS Organics™

Brand:  Acros Organics 336600050

Code : 29

Additional Details : CAS Number : 31024-26-7 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10556071

  • £156.60 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Aminopropyldimethylmethoxysilane
31024-26-7
100.0
97%
147.29
97%
96.0
≥96.0%
C6H17NOSi
MFCD01861689

Specifications

3-Aminopropyldimethylmethoxysilane
31024-26-7
96.0
Glass bottle
Clear colorless
5g
97%
147.29
97%
Authentic
100.0
1.4300 to 1.4320 (20°C, 589nm)
Liquid
≥96.0%
C6H17NOSi
MFCD01861689
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Aminopropyldimethylmethoxysilane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS