missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)phosphonic acid, 97%, ACROS Organics™

Brand:  Acros Organics 344940050

Code : 29

Additional Details : CAS Number : 13138-33-5 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10676714

  • £345.70 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

(3-Aminopropyl)phosphonic acid
96.0
97%
C3H10NO3P
139.09
13138-33-5
100.0
97%
NH2(CH2)3P(O)(OH)2
MFCD00008222

Specifications

(3-Aminopropyl)phosphonic acid
Authentic
100.0
Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
290.0° to 294.0°C
5g
97%
NH2(CH2)3P(O)(OH)2
MFCD00008222
13138-33-5
96.0
Glass bottle
Off-white to faintly greenish
Powder
97%
C3H10NO3P
139.09
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (3-Aminopropyl)phosphonic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS