missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)phosphonic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3777311
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13138-33-5
C3H10NO3P
139.09
MFCD00008222
GSZQTIFGANBTNF-UHFFFAOYSA-N
3-aminopropyl phosphonic acid, aminopropylphosphonate, 3-aminopropylphosphonicacid, unii-vm92t06vpb, 3-amino-propyl-phosphonic acid, phosphonic acid, 3-aminopropyl, vm92t06vpb, 3-phosphonopropylamine, phosphate analogue, 8, pubchem10325
97587
3-aminopropylphosphonic acid
C(CN)CP(=O)(O)O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13138-33-5
C3H10NO3P
139.09
MFCD00008222
GSZQTIFGANBTNF-UHFFFAOYSA-N
3-aminopropyl phosphonic acid, aminopropylphosphonate, 3-aminopropylphosphonicacid, unii-vm92t06vpb, 3-amino-propyl-phosphonic acid, phosphonic acid, 3-aminopropyl, vm92t06vpb, 3-phosphonopropylamine, phosphate analogue, 8, pubchem10325
97587
3-aminopropylphosphonic acid
C(CN)CP(=O)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 290.0°C to 294.0°C
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NH2(CH2)3P(O)(OH)2
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
Formula Weight 139.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (3-Aminopropyl)phosphonic acid
Show More Show Less
compliance-icons
Product Identifier
  • (3-Aminopropyl)phosphonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'