missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals

Catalog Number p-7046809
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3179-76-8
C8H21NO2Si
191.346
MFCD00039785
HXLAEGYMDGUSBD-UHFFFAOYSA-N
3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane
18511
3-[diethoxy(methyl)silyl]propan-1-amine
CCO[Si](C)(CCCN)OCC
This item is not returnable. View return policy
3179-76-8
C8H21NO2Si
191.346
MFCD00039785
HXLAEGYMDGUSBD-UHFFFAOYSA-N
3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane
18511
3-[diethoxy(methyl)silyl]propan-1-amine
CCO[Si](C)(CCCN)OCC

(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 197°C to 199°C
Melting Point <-20°C
Quantity 25 g
UN Number UN2735
Beilstein 1744264
Flash Point 85°C (185°F)
Solubility Information Soluble and reacts with water.
Formula Weight 191.35
Percent Purity 97%
Refractive Index 1.427
Sensitivity Air and moisture sensitive
Density 0.92
Chemical Name or Material (3-Aminopropyl)diethoxymethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'