missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals

Catalog Number p-58107
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
2079-89-2
0·5 C4H4O4
128.13
MFCD00078321
This item is not returnable. View return policy
2079-89-2
0·5 C4H4O4
128.13
MFCD00078321

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 170.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2NCH2CH2CN·1/2HO2CCH=CHCO2H
Quantity 5 g
Formula Weight 128.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopropionitrile fumarate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopropionitrile fumarate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'