missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylacetylene, 98%, ACROS Organics™

£37.20 - £123.33

Chemical Identifiers

CAS 54060-30-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00014779
InChI Key NNKQLUVBPJEUOR-UHFFFAOYSA-N
Synonym 3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
PubChem CID 104682
IUPAC Name 3-ethynylaniline
SMILES C#CC1=CC(=CC=C1)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10017831 SDS View Documents Acros Organics
364720050
5g Glass bottle £37.20
5g View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
10027831 SDS View Documents Acros Organics
364720250
25g Glass bottle £123.33
25g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

54060-30-9
117.15
NNKQLUVBPJEUOR-UHFFFAOYSA-N
104682
C#CC1=CC(=CC=C1)N
C8H7N
MFCD00014779
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
3-ethynylaniline

Specifications

3-Aminophenylacetylene
59°C
Glass bottle
1.04
Yellow to Brown
98%
97.5
97.5% min. (GC)
H2N(C6H4)CCH
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
C#CC1=CC(=CC=C1)N
117.15
117.15
98%
1.0400g/mL
Authentic
1.6140 to 1.616
92°C to 93°C (2.0mmHg)
5g
54060-30-9
100.0
C8H7N
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-ethynylaniline
104682
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Aminophenylacetylene

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.