missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylacetic acid, 97%, Thermo Scientific™

£42.40 - £232.00

Chemical Identifiers

CAS 14338-36-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00075058
InChI Key XUSKZLBLGHBCLD-UHFFFAOYSA-N
Synonym 3-aminophenylacetic acid, 2-3-aminophenyl acetic acid, 3-aminophenyl acetic acid, benzeneacetic acid, 3-amino, 3-aminobenzeneacetic acid, pubchem1961, 3-aminophenylaceticacid, acmc-1bovt, 3-carboxymethyl aniline, 3-amino phenylacetic acid
PubChem CID 571807
IUPAC Name 2-(3-aminophenyl)acetic acid
SMILES NC1=CC=CC(CC(O)=O)=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10122964
SDS View Documents
Thermo Scientific Acros
427610010
1g Glass bottle
£42.40
1g
View alternative products
Save Up to 
»
Estimated Shipment: 19-11-2021
Please to purchase this item. Need a web account? Register with us today!
10664175
SDS View Documents
Thermo Scientific Acros
427610100
10g Glass bottle
£232.00
10g
View alternative products
Save Up to 
»
Estimated Shipment: 26-11-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

14338-36-4
151.17
XUSKZLBLGHBCLD-UHFFFAOYSA-N
571807
NC1=CC=CC(CC(O)=O)=C1
C8H9NO2
MFCD00075058
3-aminophenylacetic acid, 2-3-aminophenyl acetic acid, 3-aminophenyl acetic acid, benzeneacetic acid, 3-amino, 3-aminobenzeneacetic acid, pubchem1961, 3-aminophenylaceticacid, acmc-1bovt, 3-carboxymethyl aniline, 3-amino phenylacetic acid
2-(3-aminophenyl)acetic acid

Specifications

3-Aminophenylacetic acid
96.0
96% min. (HPLC)
H2NC6H4CH2CO2H
MFCD00075058
Glass bottle
NC1=CC=CC(CC(O)=O)=C1
151.17
151.16
97%
147°C to 150°C
14338-36-4
100.0
C8H9NO2
Authentic
3-aminophenylacetic acid, 2-3-aminophenyl acetic acid, 3-aminophenyl acetic acid, benzeneacetic acid, 3-amino, 3-aminobenzeneacetic acid, pubchem1961, 3-aminophenylaceticacid, acmc-1bovt, 3-carboxymethyl aniline, 3-amino phenylacetic acid
XUSKZLBLGHBCLD-UHFFFAOYSA-N
2-(3-aminophenyl)acetic acid
571807
Crystalline Powder
White to Yellow
1g
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Aminophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications

RUO – Research Use Only