missing translation for 'onlineSavingsMsg'
Learn More

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Catalog Number p-4812912
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
21635-88-1
C3H7NO
73.09
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N
This item is not returnable. View return policy
21635-88-1
C3H7NO
73.09
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 99°C
Flash Point 24°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 73.09
Percent Purity 95%
Chemical Name or Material 3-Aminooxetane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminooxetane, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'